missing translation for 'onlineSavingsMsg'
Learn More

nor-Binaltorphimine, TRC

Product Code. 30330361
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330361 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330361 Supplier Toronto Research Chemicals Supplier No. TRCB38704325MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material nor-Binaltorphimine
CAS 105618-26-6
Molecular Formula C40H43N3O6
Quantity 25 mg
InChI Formula InChI=1S/C40H43N3O6/c44-25-7-5-21-13-27-39(46)15-23-24-16-40(47)28-14-22-6-8-26(45)34-30(22)38(40,10-12-43(28)18-20-3-4-20)36(49-34)32(24)41-31(23)35-37(39,29(21)33(25)48-35)9-11-42(27)17-19-1-2-19/h5-8,19-20,27-28,35-36,41,44-47H,1-4,9-18H2/t27-,28-,35+,36+,37+,38+,39-,40-/m1/s1
SMILES Oc1ccc2C[C@H]3N(CC4CC4)CC[C@@]56[C@@H](Oc1c25)c7[nH]c8[C@@H]9Oc%10c(O)ccc%11C[C@H]%12N(CC%13CC%13)CC[C@@]9(c%10%11)[C@@]%12(O)Cc8c7C[C@@]36O
IUPAC Name (1S,2S,7S,8S,12R,20R,24R,32R)-11,33-bis(cyclopropylmethyl)-19,25-dioxa-11,22,33-triazaundecacyclo[24.9.1.18,14.01,24.02,32.04,23.05,21.07,12.08,20.030,36.018,37]heptatriaconta-4(23),5(21),14(37),15,17,26,28,30(36)-octaene-2,7,17,27-tetrol
Molecular Weight (g/mol) 661.79
Formula Weight 661.3152
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.