Learn More
Invitrogen™ NOR-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534012
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Bovine, Dog, Elephant, Panda, Rabbit, Pig, Platypus (100%); Marmoset, Mouse, Rat, Bat, Hamster, Opossum, Turkey, Chicken (94%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
NOR-1, a NR4 nerve growth factor-like receptor, has been shown to affect neuronal development, T-cell apoptosis, and cancer development. NOR-1 binds to NBRE (NGFI-B response element) and regulates the pro-opiomelanocortin (POMC) gene. NOR-1 is induced rapidly but transiently by a variety of stimuli. Downregulation of NOR-1 and other NGFI-B family members is correlated with drug addiction-prone behavior of Lewis rats. Three human protein isoforms have been documented: NOR-1, NOR-1beta, and NOR-1 3' variant.
Specifications
| NOR-1 | |
| Polyclonal | |
| Unconjugated | |
| NR4A3 | |
| AI573420; CHN; chondrosarcoma, extraskeletal myxoid, fused to EWS; CSMF; MINOR; mitogen-induced nuclear orphan receptor; Neuron-derived orphan receptor 1; neuron-derived orphan receptor 1/2; Nor1; NOR-1; NOR-2; Nr4a3; Nuclear hormone receptor NOR-1; nuclear hormone receptor NOR-1/NOR-2; Nuclear receptor subfamily 4 group A member 3; nuclear receptor subfamily 4, group A, member 3; Orphan nuclear receptor TEC; TEC; TEC receptor; Translocated in extraskeletal chondrosarcoma | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100344785, 100684628, 397479, 528877, 8013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q92570 | |
| NR4A3 | |
| Synthetic 17 amino acid peptide from internal region of human NR4A3. | |
| 50 μg | |
| Primary | |
| Human, Canine, Rabbit, Bovine, Pig | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.