missing translation for 'onlineSavingsMsg'
Learn More

Nope/IGDCC4 Antibody (2516A) [Janelia Fluor™ 669], Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals FAB10345JF669

Product Code. 30158739

  • £426.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Nope/IGDCC4 Monoclonal antibody specifically detects Nope/IGDCC4 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nope/IGDCC4
Monoclonal
Janelia Fluor 669
DDM36, hDDM36, immunoglobulin superfamily DCC subclass member 4, immunoglobulin superfamily, DCC subclass, member 4, KIAA1628, likely ortholog of mouse neighbor of Punc E11, LOC57722, Neighbor of punc e11, NOPEFLJ42051, Protein DDM36
Chinese hamster ovary cell line CHO-derived recombinant human Nope/IGDCC4, Met1-Thr957, Accession # Q8TDY8
RUO
57722
Store at 4°C in the dark.
IgG
Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF
2516A
50mM Sodium Borate
Rabbit
0.1 mL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.