missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOP58 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554321
This item is not returnable.
View return policy
Description
Immunogen sequence: LDKELNNYIM RCREWYGWHF PELGKIISDN LTYCKCLQKV GDRKNYASAK LSELLPEEVE AEVKAAAEIS MGTEVSEEDI CNILHLCTQV IEISEYRTQL YEYLQNRMMA IAPNVTVMVG ELVGARLIAH AGSLL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Nop5p, or Nop58p, is localized in the nucleolus and required for pre-18S rRNA processing in Saccharomyces cerevisiae.
Specifications
| NOP58 | |
| Polyclonal | |
| Unconjugated | |
| NOP58 | |
| HSPC120; MSSP; Nap65; Nol5; NOP5; NOP5/NOP58; Nop58; NOP58 ribonucleoprotein; NOP58 ribonucleoprotein homolog; Nopp140 associated protein; nopp140-associated protein of 65 kDa; Nucleolar protein 5; nucleolar protein 58; nucleolar protein NOP5/NOP58; nucleolar protein NOP58; O6108; RNA-processing protein NOP58; SIK; SIK similar protein; SIK-similar protein; YOR310C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51602, 55989, 60373 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6DFW4, Q9QZ86, Q9Y2X3 | |
| NOP58 | |
| Recombinant protein corresponding to Human NOP58. Recombinant protein control fragment (Product #RP-91831). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction