missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP56 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-46847
This item is not returnable.
View return policy
Description
NOP56 Polyclonal specifically detects NOP56 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| NOP56 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| NOL5A, NOP56 ribonucleoprotein homolog (yeast), nucleolar protein 56, Nucleolar protein 5A, nucleolar protein 5A (56kD with KKE/D repeat), nucleolar protein 5A (56kDa with KKE/D repeat) | |
| Rabbit | |
| 66 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Gorilla.. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| O00567 | |
| NOP56 | |
| The immunogen for this product maps to a region between residue 494 and 544 of human NOP56 ribonucleoprotein homolog using the numbering given in entry NP_006383.2 (GeneID 10528). | |
| Affinity Purified | |
| RUO | |
| 10528 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction