missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP56 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35887-20ul
This item is not returnable.
View return policy
Description
NOP56 Polyclonal antibody specifically detects NOP56 in Human samples. It is validated for ELISA,Western Blot
Specifications
| NOP56 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| NOL5A, NOP56 ribonucleoprotein homolog (yeast), nucleolar protein 56, Nucleolar protein 5A, nucleolar protein 5A (56kD with KKE/D repeat), nucleolar protein 5A (56kDa with KKE/D repeat) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human NOP56 (NP_006383.2).,, Sequence:, MVLLHVLFEHAVGYALLALKEVEEISLLQPQVEESVLNLGKFHSIVRLVAFCPFASSQVALENANAVSEGVVHEDLRLLLETHLPSKKKKVLLGVGDPKIGAAIQEELGYNCQTGGVIAEILRGVRLHFHNLVKGLTDLSACKAQLGLGHSYSRAKVKFN | |
| 20 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10528 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction