Learn More
Invitrogen™ NOP16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589637
Description
Immunogen sequence: MPKAKGKTRR QKFGYSVNRK RLNRNARRKA APRIECSHIR HAWDHAKSVR QNLAEMGLAV DPNRAVPLRK RKVKAMEVDI EERPKELVRK PYVLNDLEAE ASLPEKKGNT LSRDLIDYVR YMVENHGEDY KAMARDEKNY YQDTPKQIRS KINVYKRFYP AEWQDFLDSL QKRKMEVE; Positive Samples: HT-29, A-549, A-431, MCF-7, HeLa, Mouse thymus, Rat spleen; Cellular Location: Nucleus, nucleolus.
NOP16 is transcriptionally regulated by c-Myc (MYC), upregulated in breast cancer, and overexpression is associated with poor patient survival (Butt et al., 2008).
Specifications
| NOP16 | |
| Polyclonal | |
| Unconjugated | |
| Nop16 | |
| AA409471; CGI-117; D13Wsu177e; HBV pre-S2 trans-regulated protein 3; HSPC111; HSPC185; hypothetical protein HSPC111; Nop16; NOP16 nucleolar protein; NOP16 nucleolar protein homolog; nucleolar protein 16; nucleolar protein 16 homolog; nucleolar protein 16-like; Protein ZH1; RGD1305727 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 28126, 306768, 51491 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q1RP77, Q9CPT5, Q9Y3C1 | |
| Nop16 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-178 of human NOP16 (NP_0574752). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.