missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56381
This item is not returnable.
View return policy
Description
NOP10 Polyclonal specifically detects NOP10 in Human samples. It is validated for Western Blot.
Specifications
| NOP10 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| H/ACA ribonucleoprotein complex subunit 3, homolog of yeast Nop10p, MGC70651, NOLA3Nucleolar protein family A member 3, NOP10 ribonucleoprotein homolog (yeast), NOP10P, Nucleolar protein 10, nucleolar protein family A, member 3 (H/ACA small nucleolar RNPs), snoRNP protein NOP10 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| This product is specific to Subunit or Isoform: 3. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NPE3 | |
| NOP10 | |
| Synthetic peptides corresponding to NOLA3 The peptide sequence was selected from the middle region of NOLA3. Peptide sequence MFLQYYLNEQGDRVYTLKKFDPMGQQTCSAHPARFSPDDKYSRHRITIKK. | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 55505 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction