missing translation for 'onlineSavingsMsg'
Learn More

Nonyl Glycidyl Ether, TRC

Product Code. 30470087
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470087 50 mg -
30474144 500 mg -
30469701 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470087 Supplier Toronto Research Chemicals Supplier No. TRCN33726550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10580-65-1
Molecular Formula C12 H24 O2
Molecular Weight (g/mol) 200.318
Synonym NSC 291916, 3-Nonyloxy-1-propene Oxide, 2-[(Nonyloxy)methyl]oxirane, 1,2-Epoxy-3-(nonyloxy)propane, [(Nonyloxy)methyl]oxirane
IUPAC Name 2-(nonoxymethyl)oxirane
SMILES CCCCCCCCCOCC1CO1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H24O2/c1-2-3-4-5-6-7-8-9-13-10-12-11-14-12/h12H,2-11H2,1H3
Formula Weight 200.178
Chemical Name or Material Nonyl Glycidyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.