missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ Nonwoven Polyester Lab-Tips™ Swabs

Nonwoven Polyester Swabs are available with small and long handles.

Brand:  Berkshire™ LTN1465.10

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12886804

  • £470.00 / Pack of 1000
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

  • Double layer, hydroentangled polyester over flexible paddle
  • Soft and non abrasive
  • Thermally bonded head
  • Packaged in an ISO class 4 (Class 10) cleanroom
Specifications

Specifications

RoHS 2002/05 and REACH
6.38 in.
Swab
16.2cm
14.5cm
1.7cm
0.24 in.
Polypropylene
Non-woven Polyester
0.157 in.
Cleaning of sensitive equipment, applying lubricants and other liquids, cleaning validation
0.31cm
5.71 in.
0.67 in.
12.7mm
Non-contaminating
Round
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers