Learn More
Invitrogen™ Nonspecific Cytotoxic Cell Monoclonal Antibody (5C.6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1760
Description
MA1-760 detects nonspecific cytotoxic cells (NCC) from catfish and tilapia tissues and also detects natural killer/lymphokine activated killer (NK/LAK) cells from rat, bovine, equine, canine and porcine tissues. MA1-760 has been successfully used in Western blot, cytotoxic inhibition studies with NCC/NK/LAK cells, FACS, immunoprecipitation, panning, and ELISA procedures. By Western blot, this antibody detects a 32 kDa protein representing nonspecific cytotoxic cell receptor protein. In cytotoxicity assays, this antibody is used under saturating conditions to inhibit target cell conjugation. The MA1-760 immunogen is purified NCC cells from catfish.
Non-specific cytotoxic cells (NCCs) in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. One such property is that NCC/natural killer (NK)/lymphokine activated killer (LAK) cells express spontaneous, non-major histocompatibility complex restricted cytotoxic activity. NCC and LAK lyse a variety of transformed murine and human B-cell, T-cell and myeloid targets. A 32 kDa membrane protein [non-specific cytotoxic cell receptor protein (NCCRP-1)] expressed by NCC and certain mammalian NK/LAK cells mediates this cytotoxicity. NCCRP-1 is evolutionarily conserved and is found in species ranging from marine and freshwater teleosts to higher mammals.
Specifications
| Nonspecific Cytotoxic Cell | |
| Monoclonal | |
| 0.73 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| Mouse | |
| Purified | |
| RUO | |
| Multi-species | |
| Antibody | |
| IgM |
| ELISA, Flow Cytometry, Inhibition Assays, Immunoprecipitation, Western Blot | |
| 5C.6 | |
| Unconjugated | |
| Natural Killer Cell; NCC/NK | |
| Purified NCC cells from catfish. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.