missing translation for 'onlineSavingsMsg'
Learn More

NONO Antibody (OTI3F8), DyLight 594, Novus Biologicals™

Product Code. 18400795 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400795 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400795 Supplier Novus Biologicals Supplier No. NBP273033DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NONO Monoclonal antibody specifically detects NONO in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NONO
Applications Western Blot, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone OTI3F8
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias NMT5552 kDa subunit, non-POU domain containing, octamer-binding, NRB54non-POU-domain-containing, octamer-binding, p54(nrb)
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 184-385 of human NONO (NP_031389) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4841
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.