missing translation for 'onlineSavingsMsg'
Learn More

Nonivamide, MedChemExpress

Product Code. 16437376
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
5 g
Unit Size:
100mg
500mg
5g
This item is not returnable. View return policy

Product Code. 16437376

Brand: Medchem Express HY17568

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nonivamide is a agonist, which exhibits 4d-EC50 value of 5.1 mg/L in static toxicity tests.

Nonivamide is a agonist, which exhibits 4d-EC50 value of 5.1 mg/L in static toxicity tests.

Specifications

Chemical Name or Material Nonivamide
CAS 2444-46-4
Color White
Purity Grade Notes Research
Molecular Formula C17H27NO3
Quantity 100 mg
Synonym Pelargonic acid vanillylamide Nonanoic acid vanillylamide Pseudocapsaicin
Solubility Information DMSO : 100 mg/mL (340.83 mM; Need ultrasonic)
SMILES CCCCCCCCC(NCC1=CC=C(O)C(OC)=C1)=O
Molecular Weight (g/mol) 293.4
Formula Weight 293.4
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.