missing translation for 'onlineSavingsMsg'
Learn More

Melford Nonidet P40 Substitute

Product Code. 30018595
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018595 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018595 Supplier Melford Supplier No. N59000500.0
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratory reagent grade.

Laboratory reagent grade. Nonidet P-40 is a nonionic, non-denaturing detergent.

Specifications

Chemical Name or Material Nonidet P-40 Substitute
CAS 9002-93-1
Quantity 500 mL
Molecular Formula C16H26O2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.