Learn More
MedChemExpress Nonapeptide-1 acetate salt
Description
Nonapeptide-1 acetate salt is a competitive α-MSH antagonist that potently inhibits intracellular cAMP and melanosome dispersion induced by α-MSH in melanocytes (IC50: 2.5 nM and 11 nM, respectively). Nonapeptide-1 acetate salt inhibits melanin synthesis, and can be used in the research of skin pigmentation and regulation of steroid production in the adrenal gland, skin cancer.
Specifications
Specifications
| Purity | 96.64% |
| Content And Storage | Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 1266.56 g/mol |
| Product Type | Peptides |
| Quantity | 1 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 50 mg/mL (39.48 mM∣Need ultrasonic) |
| Synonym | Melanostatine-5 acetate salt |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.