missing translation for 'onlineSavingsMsg'
Learn More

Nonanal, TRC

Product Code. 30135980
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135980 10 g 10g
30137484 50 g 50g
30009912 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135980 Supplier Toronto Research Chemicals Supplier No. TRCN64933010G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-19-6
Molecular Formula C9 H18 O
Molecular Weight (g/mol) 142.24
IUPAC Name nonanal
SMILES CCCCCCCCC=O

Specifications

Quantity 10 g
Synonym Nonanal, Nonanaldehyde (7CI), Aldehyde C 09, Aldehyde C 9, NSC 5518, Nonaldehyde, Nonanoic aldehyde, Nonylaldehyde, Nonylic aldehyde, Pelargonaldehyde, Pelargonic aldehyde, n-Nonanal, n-Nonylaldehyde
InChI Formula InChI=1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h9H,2-8H2,1H3
Formula Weight 142.1358
Chemical Name or Material Pelargonaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.