missing translation for 'onlineSavingsMsg'
Learn More

Nonafluoropentanoic Acid, TRC

Product Code. 17903955
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903955 250 mg 250mg
17953605 100 mg 100mg
17970905 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17903955 Supplier Toronto Research Chemicals Supplier No. TRCN649515250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2706-90-3
Molecular Formula C5 H F9 O2
Molecular Weight (g/mol) 264.05
Synonym Pentanoic acid, nonafluoro- (9CI), Valeric acid, nonafluoro- (6CI,8CI), 2,2,3,3,4,4,5,5,5-Nonafluoropentanoic acid, NSC 18404, Nonafluoropentanoic acid, Nonafluorovaleric acid, PFPeA, Perfluoropentanoic acid, Perfluorovaleric acid
IUPAC Name 2,2,3,3,4,4,5,5,5-nonafluoropentanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5HF9O2/c6-2(7,1(15)16)3(8,9)4(10,11)5(12,13)14/h(H,15,16)
Formula Weight 263.9833
Percent Purity >95%
Chemical Name or Material Perfluoropentanoic acid
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.