missing translation for 'onlineSavingsMsg'
Learn More

Nonafluoropentanoic acid, 97%

Product Code. p-3737084 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10730591

Brand: Thermo Scientific Acros 309290050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2706-90-3 | C5HF9O2 | 264.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2706-90-3
Molecular Formula C5HF9O2
Molecular Weight (g/mol) 264.05
MDL Number MFCD00040211
InChI Key CXZGQIAOTKWCDB-UHFFFAOYSA-N
Synonym perfluoropentanoic acid, perfluorovaleric acid, nonafluoropentanoic acid, nonafluorovaleric acid, pentanoic acid, nonafluoro, nonafluoro-1-pentanoic acid, valeric acid, nonafluoro, n-perfluoropentanoic acid, perfluoro-n-pentanoic acid, 2,2,3,3,4,4,5,5,5-nonafluoro-pentanoic acid
PubChem CID 75921
ChEBI CHEBI:83491
IUPAC Name 2,2,3,3,4,4,5,5,5-nonafluoropentanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.7100g/mL
Boiling Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CF3(CF2)3CO2H
Refractive Index 1.2940 to 1.297
Quantity 5 mL
Specific Gravity 1.71
Formula Weight 264.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Nonafluoropentanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Nonafluoropentanoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nonafluoropentanoicacid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.