missing translation for 'onlineSavingsMsg'
Learn More

Nonadecane, TRC

Product Code. 30466094
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466094 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466094 Supplier Toronto Research Chemicals Supplier No. TRCN6489501G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Nonadecane
CAS 629-92-5
Molecular Formula C19 H40
Quantity 1 g
Synonym Nonadecane, NSC 77136, n-Nonadecane
InChI Formula InChI=1S/C19H40/c1-3-5-7-9-11-13-15-17-19-18-16-14-12-10-8-6-4-2/h3-19H2,1-2H3
SMILES CCCCCCCCCCCCCCCCCCC
IUPAC Name nonadecane
Molecular Weight (g/mol) 268.52
Formula Weight 268.313
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.