missing translation for 'onlineSavingsMsg'
Learn More

Nonadecane, 99%

Product Code. 10419440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10419440 25 g Glass bottle 25g
10732515 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10419440 Supplier Thermo Scientific Acros Supplier No. 129100250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 629-92-5 | C19H40 | 268.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-92-5
Molecular Formula C19H40
Molecular Weight (g/mol) 268.51
MDL Number MFCD00009012
InChI Key LQERIDTXQFOHKA-UHFFFAOYSA-N
Synonym n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl
PubChem CID 12401
ChEBI CHEBI:32927
IUPAC Name nonadecane
SMILES CCCCCCCCCCCCCCCCCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 32.0°C to 34.0°C
Color White
Boiling Point 330.0°C
Flash Point 168°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)17CH3
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Viscosity 3.6 mPa.s (40°C)
Formula Weight 268.51
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Nonadecane
Show More Show Less
compliance-icons
Product Identifier
  • Nonadecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.