Learn More
Greiner Bio-One Non-treated 384-well Microplates
Product Code. 10527071
Change view
Click to view available options
Color:
Black
Clear
Quantity:
100/Cs.
40/Cs.
80/Pk.
Lid:
Without Lid
Total Volume (Metric) Well:
28 μL
131 μL
Unit Size:
Pack of 100
Pack of 40
Pack of 80
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity Lid Total Volume (Metric) Well unitSize
10527071 Black 40/Cs. Without Lid 131 μL Pack of 40
10446152 Clear 80/Pk. - 131 μL Pack of 80
10060202 Clear 100/Cs. Without Lid 131 μL Pack of 100
10737112 Clear 40/Cs. Without Lid 28 μL Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10527071 Supplier Greiner Bio-One Supplier No. 781096
Manufactured without surface treatment to create low biomolecular binding. Greiner Bio One™ Non-treated 384-Well Assay Microplates are ideal for sensitive biochemical screening applications.
  • Footprint compatible with automated systems
  • Can be traced to production through defined LOT number system
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Quadrupled the number of wells (from 96)
  • Well-to-well spacing: 4.5 mm
  • Rounded square well profile, providing flexible working volume of 10 μl to 130 μl and reduced wicking and bubbling
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN ISO 9001
Format Standard
Lid Without Lid
Array 16 x 24
Binding Type Medium Binding
Color Black
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 131 μL
Well Shape Flat Bottom (F-well)
No. per Pack 40
Quantity 40/Cs.