missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Non-species specific IgG2a Isotype Control (IGG2a/6723) [DyLight 650], Novus Biologicals™

Product Code. 30062794 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30062794 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30062794 Supplier Novus Biologicals™ Supplier No. NBP327042C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG2a Monoclonal antibody specifically detects IgG2a in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2a
Applications Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence
Classification Monoclonal
Clone IGG2a/6723
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Host Species Mouse
Immunogen Recombinant full-length human IgG2a protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.