missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
non-muscle Myosin IIA Polyclonal specifically detects non-muscle Myosin IIA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | non-muscle Myosin IIA |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | Cellular myosin heavy chain, type A, DFNA17, EPSTS, FTNSNMMHC-IIA, MGC104539, MHANMMHC II-a, MYH9 variant protein, Myosin heavy chain 9, Myosin heavy chain, non-muscle IIa, myosin, heavy chain 9, non-muscle, myosin, heavy polypeptide 9, non-muscle, myosin-9, NMHC-II-A, NMMHCA, NMMHC-A, Non-muscle myosin heavy chain A, nonmuscle myosin heavy chain II-A, Non-muscle myosin heavy chain IIa, non-muscle myosin heavy polypeptide 9 |
| Gene Symbols | MYH9 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:REQEVNILKKTLEEEAKTHEAQIQEMRQKHSQAVEELAEQLEQTKRVKANLEKAKQTLENERGELANEVKVLLQGKGDSEHKRKKVEAQLQELQVKFNEGERVRTELADKVTKLQVELDNVTGLLSQSDSKSSKLTKDF |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?