missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QIAGEN Non-Indicating FTA™ Micro Card
Description
FTA technology consists of two distinct chemistries, both of which have the ability to lyse cells on contact, denature proteins, and protect DNA from degradation. FTA chemistry contains chemical denaturants and a free radical scavenger. With FTA, the DNA remains tightly bound while proteins and inhibitors are washed from the matrix.
- One sample area per card
- Improves process efficiency by enabling direct amplification for STR analysis
- Non-indicating cards for colored samples, like blood
- Immediate stabilization and protection of nucleic acids due to FTA technology
- Different card options to support fast manual or automated processing
- Long term archiving of samples at room temperature due to FTA card technology
Specifications
Specifications
| Format | Card |
| Sample Type | Blood |
| For Use With (Application) | Forensics, Transgenic Identification, HLA Typing/Transfusion Medicine |
| No. of Samples | 1 |
| Product Type | Non-Indicating FTA Micro Card |
| Quantity | 125 μL |
| Sample Volume | 125μL |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?