missing translation for 'onlineSavingsMsg'
Learn More

QIAGEN QIAcard FTA Classic

Product Code. 11310854
Change view
Click to view available options
Quantity:
100 Pk.
25 Pk.
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11310854 25 Pk. Pack of 25
10223352 100 Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310854 Supplier QIAGEN Supplier No. WB120305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Classic with 4 sample areas per card.

  • QIAcard FTA Classic format with 4 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
  • FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.

Specifications

Description Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Classic Cards.
Format Card
Sample Type Blood
For Use With (Equipment) Direct STR amplification
For Use With (Application) Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
No. of Samples 4
Product Type FTA Classic Card
Quantity 25 Pk.
Sample Volume 125 mL
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.