missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Non-human Primate IL-23 ELISA Kit

Product Code. 18631857 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit (1 Plate)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18631857

Brand: Novus Biologicals™ NBP311730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 10 to 3160 pg/mL
Assay Sensitivity 5 pg/mL
Immunoassay Kit Format ELISA (Standard)
Product Type ELISA Kit
Sample Type Serum, Plasma, Cell Culture Supernatant
Target Species Non-human Primate
For Use With (Application) ELISA
Gene ID (Entrez) 51561
Gene Symbol IL23A
Quantity 1 Kit (1 Plate)
Research Discipline Cancer, Cytokine Research, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12 B, IL-23, IL-23 subunit alpha, IL-23 A, IL-23-A, IL-23p19, IL23P19P19, interleukin 12, p40, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-12 beta chain, interleukin-12 subunit beta, Interleukin-23 subunit p19, interleukin-six, G-CSF related factor, JKA3 induced upon T-cell activation, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, P19, SGRF
Target Non-human Primate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.