missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Non-human Primate IFN-alpha ELISA Kit

Product Code. 18374282 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit (1 Plate)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374282 1 Kit (1 Plate) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374282 Supplier Novus Biologicals™ Supplier No. NBP311725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 5 - 4000 pg/mL
Assay Sensitivity 2 pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 3440
Gene Symbol IFNA2
Quantity 1 Kit (1 Plate)
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym IFNA, IFNA2, IFNA2b, IFN-alphaA, INFA2, interferon alpha 2
Target Non-human Primate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.