missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Non-human Primate Apolipoprotein H/ApoH ELISA Kit

Product Code. 18652178 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit (1 Plate)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18652178

Brand: Novus Biologicals™ NBP311736

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.02 to 15 ng/mL
Assay Sensitivity 0.02 ng/mL
Immunoassay Kit Format ELISA (Standard)
Product Type ELISA Kit
Sample Type Serum, Plasma, Cell Culture Supernatant
Target Species Non-human Primate
For Use With (Application) ELISA
Gene ID (Entrez) 350
Gene Symbol APOH
Quantity 1 Kit (1 Plate)
Research Discipline Cholesterol Metabolism, Lipid and Metabolism
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG
Target Non-human Primate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.