Learn More
Greiner Bio-One Non-binding Surface 384-well Microplates
Product Code. 11897311
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 40
Product Code. Color unitSize
11897311 Black Pack of 40
1 options
Product Code. 11897311 Supplier Greiner Bio-One Supplier No. 784900
Only null left
Add to basket
Add to basket
Characterized by low protein, peptide, DNA and RNA binding properties, the non-binding surface increases assay sensitivity by reducing background, improving signal-to-noise ration. Greiner Bio One™Non-Binding Surface 384-Well Microplates remain stable under common assay conditions and does not degrade during short-term storage.
Characteristic features of the non-binding surface are: 
  • Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA) 
  • Long-term surface stability without degradation or leaching 
  • Higher assay sensitivity with reduced background
Advantages: significantly increase assay sensitivity--reducing background--improving signal-to-noise ratio 
  • Free of detectable DNase, RNase, human DNA 
  • Non-pyrogenic 
  • Manufactured under DIN ISO 9001 guidelines 
  • Can be traced all the way back to production through a defined LOT number system 
  • Well profile: Small Volume™ 
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Certifications/Compliance DIN ISO 9001
Format Standard
Array 16 x 24
Binding Type Non Binding
Color Black
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
Surface Treatment Protein-Repellent Non-binding
Total Volume (Metric) Well 28 μL
Well Shape Flat Bottom (F-well)
No. per Pack 40
Quantity 40/Cs.