missing translation for 'onlineSavingsMsg'
Learn More

Nomilin (~85%), TRC

Product Code. 30481340
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481340 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481340 Supplier Toronto Research Chemicals Supplier No. TRCN63595350MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nomilin
CAS 1063-77-0
Molecular Formula C28 H34 O9
Quantity 50 mg
Synonym Oxireno[4,4a]-2-benzopyrano[6,5-g][2]benzoxepin-3,5,9(3aH,4bH,6H)-trione','11-(acetyloxy)-1-(3-furanyl)decahydro-4b,7,7,11a,13a-pentamethyl-','(1S,3aS,4aR,4bR,6aR,11S,11aR,11bR,13aS)-, Nomilin (6CI,7CI), Obacunoic acid','1-(acetyloxy)-1,2-dihydro-','ε-lactone, Oxireno[4,4a]-2-benzopyrano[6,5-g][2]benzoxepin-3,5,9(3aH,4bH,6H)-trione','1-(3-furyl)decahydro-11-hydroxy-4b,7,7,11a,13a-pentamethyl-','acetate (8CI), (1S,3aS,4aR,4bR,6aR,11S,11aR,11bR,13aS)-11-(Acetyloxy)-1-(3-furanyl)decahydro-4b,7,7,11a,13a-pentamethyloxireno[4,4a]-2-benzopyrano[6,5-g][2]benzoxepin-3,5,9(3aH,4bH,6H)-trione, NSC 297134, Oxireno[4,4a]-2-benzopyrano[6,5-g][2]benzoxepin-3,5,9(3aH,4bH,6H)-trione','11-(acetyloxy)-1-(3-furanyl)decahydro-4b,7,7,11a,13a-pentamethyl-','[1S-(1α,3aα,4aS*,4bβ,6aα,11α,11aβ,11bα,13aα)]-
InChI Formula InChI=1S/C28H34O9/c1-14(29)34-19-12-20(31)36-24(2,3)17-11-18(30)27(6)16(26(17,19)5)7-9-25(4)21(15-8-10-33-13-15)35-23(32)22-28(25,27)37-22/h8,10,13,16-17,19,21-22H,7,9,11-12H2,1-6H3/t16-,17+,19+,21+,22-,25+,26-,27+,28-/m1/s1
SMILES CC(=O)O[C@H]1CC(=O)OC(C)(C)[C@@H]2CC(=O)[C@]3(C)[C@H](CC[C@@]4(C)[C@@H](OC(=O)[C@H]5O[C@@]345)c6cocc6)[C@@]12C
Molecular Weight (g/mol) 514.56
Formula Weight 514.2203
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.