missing translation for 'onlineSavingsMsg'
Learn More

Nomifensine, MedChemExpress

Product Code. 16448616
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448616 100 mg 100mg
16444547 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448616 Supplier Medchem Express Supplier No. HYB1110
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Nomifensine is a norepinephrine-dopamine reuptake inhibitor, increases the amount of synaptic norepinephrine and dopamine available to receptors by blocking the dopamine and norepinephrine reuptake transporters.

Nomifensine is a norepinephrine-dopamine reuptake inhibitor, increases the amount of synaptic norepinephrine and dopamine available to receptors by blocking the dopamine and norepinephrine reuptake transporters.

Specifications

Chemical Name or Material Nomifensine
CAS 24526-64-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H18N2
Quantity 100 mg
Synonym (±)-Nomifensin
Solubility Information DMSO : ≥ 100 mg/mL (419.59 mM)
SMILES NC1=CC=CC2=C1CN(C)CC2C3=CC=CC=C3
Molecular Weight (g/mol) 238.33
Formula Weight 238.33
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.