missing translation for 'onlineSavingsMsg'
Learn More

N^w,N^w'-Di-Boc-Nalpha-Fmoc-L-arginine, 95%

Product Code. 15410669
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15410669

Brand: Thermo Scientific Alfa Aesar H63784.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143824-77-5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143824-77-5
MDL Number MFCD00237008
Synonym Fmoc-Arg(Boc)2-OH; (S,E)-5-[1,2-Bis(tert-butoxycarbonyl)guanidino]-2-(Fmoc-amino)pentanoic acid

Specifications

Quantity 5 g
Formula Weight 596.67
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Nomega,Nomega'-Di-Boc-Nalpha-Fmoc-L-arginine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.