missing translation for 'onlineSavingsMsg'
Learn More

Nomega-(4-Methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine monohydrate, 95%, Thermo Scientific™

Catalog Number 15458456
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
80745-10-4
C16H26N4O5S
386.467
MFCD01862361
FUSAEZSSVDNYPO-LBPRGKRZSA-N
h-arg mtr-oh, h-arg mtr-oh.1/2h2o, s-2-amino-5-3-4-methoxy-2,3,6-trimethylphenyl sulfonyl guanidino pentanoic acid, l-arginyl-4-methoxy-2,3,6-trimethylbenzenesulfonyl, n'-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, nomega-4-methoxy-2,3,6-trimethylbenzenesulfonyl-d-arginine, nomega-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, amino acid series, arginine mtr-oh, n pound inverted question mark 4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine
7408178
(2S)-2-amino-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]pentanoic acid
CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)N)N)C)C)OC
This item is not returnable. View return policy
80745-10-4
C16H26N4O5S
386.467
MFCD01862361
FUSAEZSSVDNYPO-LBPRGKRZSA-N
h-arg mtr-oh, h-arg mtr-oh.1/2h2o, s-2-amino-5-3-4-methoxy-2,3,6-trimethylphenyl sulfonyl guanidino pentanoic acid, l-arginyl-4-methoxy-2,3,6-trimethylbenzenesulfonyl, n'-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, nomega-4-methoxy-2,3,6-trimethylbenzenesulfonyl-d-arginine, nomega-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, amino acid series, arginine mtr-oh, n pound inverted question mark 4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine
7408178
(2S)-2-amino-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]pentanoic acid
CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)N)N)C)C)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 404.48
Percent Purity 95%
Quantity 5 g
Sensitivity Moisture sensitive
Physical Form Powder
Chemical Name or Material Nomega-(4-Methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'