missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOM1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18488V
This item is not returnable.
View return policy
Description
NOM1 Polyclonal antibody specifically detects NOM1 in Human samples. It is validated for Western Blot
Specifications
| NOM1 | |
| Polyclonal | |
| Western Blot | |
| C7orf3, chromosome 7 open reading frame 3, FLJ16401, nucleolar MIF4G domain-containing protein 1, nucleolar protein with MIF4G domain 1, SGD1, SGD1 homolog | |
| Recognizes region between residues Between between 200 and 250.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64434 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction