missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOLC1 Recombinant Rabbit Monoclonal Antibody (24GB490)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | NOLC1 |
|---|---|
| Clone | 24GB490 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276525
![]() |
Invitrogen™
MA557127 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274863
|
Invitrogen™
MA557128 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NOLC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Related to nucleologenesis, may play a role in the maintenance of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It has intrinsic GTPase and ATPase activities. May play an important role in transcription catalyzed by RNA polymerase I.Specifications
| NOLC1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| NOLC1 | |
| 140 kDa nucleolar phosphoprotein; HCV NS5A trans-regulated protein 13; HCV NS5A-transactivated protein 13; Hepatitis C virus NS5A-transactivated protein 13; KIAA0035; NOLC1; nolc1.L; nopp130; nopp140; NS5ATP13; Nucleolar 130 kDa protein; nucleolar and coiled-body phosphoprotein 1; nucleolar and coiled-body phosphoprotein 1 L homeolog; nucleolar and coiled-body phosphprotein 1; nucleolar phosphoprotein; nucleolar phosphoprotein 140; nucleolar phosphoprotein p130; nucleolar protein p130; P130; XELAEV_18034881mg; xNopp180 | |
| NOLC1 | |
| Primary | |
| Affinity chromatography |
| 24GB490 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14978 | |
| 9221 | |
| A synthesized peptide derived from human NOLC1 (aa 51-84). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title