missing translation for 'onlineSavingsMsg'
Learn More

NOLC1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18402963 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402963 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402963 Supplier Novus Biologicals Supplier No. NBP122981DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOLC1 Polyclonal antibody specifically detects NOLC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOLC1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 140 kDa nucleolar phosphoprotein, HCV NS5A trans-regulated protein 13, HCV NS5A-transactivated protein 13, Hepatitis C virus NS5A-transactivated protein 13, KIAA0035NS5ATP13, NOPP130, NOPP140, Nucleolar 130 kDa protein, nucleolar and coiled-body phosphoprotein 1, nucleolar and coiled-body phosphprotein 1, Nucleolar phosphoprotein p130, nucleolar protein p130, P130
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human nucleolar and coiled-body phosphoprotein 1 using the numbering given in entry NP_004732.2 (GeneID 9221).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9221
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.