missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Cell Cycle and Replication |
| Target | NOLC1 |
| Synonym | 140 kDa nucleolar phosphoprotein, HCV NS5A trans-regulated protein 13, HCV NS5A-transactivated protein 13, Hepatitis C virus NS5A-transactivated protein 13, KIAA0035NS5ATP13, NOPP130, NOPP140, Nucleolar 130 kDa protein, nucleolar and coiled-body phosphoprotein 1, nucleolar and coiled-body phosphprotein 1, Nucleolar phosphoprotein p130, nucleolar protein p130, P130 |
| Gene Symbol | NOLC1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?