missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16083-1-AP-150UL
This item is not returnable.
View return policy
Description
NOL9 is a polynucleotide 5′-kinase involved in rRNA processing. The kinase activity is required for the processing of the 32S precursor into 5.8S and 28S rRNAs, more specifically for the generation of the major 5.8S(S) form. In vitro, it has both DNA and RNA 5′-kinase activities and probably binds RNA.Specifications
| NOL9 | |
| Polyclonal | |
| Unconjugated | |
| NOL9 | |
| NOL9, nucleolar protein 9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79707 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5SY16 | |
| NOL9 | |
| NOL9 Fusion Protein Ag9082 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction