missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOL8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551027
This item is not returnable.
View return policy
Description
NOL8 Polyclonal Antibody detects endogenous levels of NOL8 protein.
NOL8 binds Ras-related GTP-binding proteins and plays a role in cell growth.
Specifications
| NOL8 | |
| Polyclonal | |
| Unconjugated | |
| NOL8 | |
| 4921532D18Rik; 5730412B09Rik; bA62C3.3; bA62C3.4; C9orf34; NOL8; NOP132; Nucleolar protein 8; nucleolar protein Nop132 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55035 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q76FK4 | |
| NOL8 | |
| Synthesized peptide derived from the C-terminal region of human Nop132. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction