missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15706-1-AP-150UL
This item is not returnable.
View return policy
Description
The nucleolus is a dense subnuclear membraneless organelle that assembles around clusters of rRNA genes and functions in ribosome biogenesis. This gene encodes a nucleolar RNA-associated protein that is highly conserved between species. RNase treatment of permeabilized cells indicates that the nucleolar localization is RNA dependent. Further studies suggest that the protein is associated with ribosome biogenesis through an interaction with pre-rRNA primary transcripts. Alternative splicing has been observed at this locus and two splice variants encoding distinct isoforms have been identified.Specifications
| NOL6 | |
| Polyclonal | |
| Unconjugated | |
| NOL6 | |
| NOL6, NRAP, Nucleolar protein 6, UTP22 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 230082, 313167, 65083 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8R5K4, Q9H6R4 | |
| Nol6 | |
| NOL6 Fusion Protein Ag8378 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction