missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568256
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
NOL4 gene ontology (GO) annotation includes RNA binding and nucleolus.
Specifications
| NOL4 | |
| Polyclonal | |
| Unconjugated | |
| Nol4 | |
| 1700013J13Rik; 4930568N03Rik; cancer/testis antigen 125; CT125; Gm1262; HRIHFB2255; Nol4; NOLP; nucleolar localized protein; nucleolar protein 4; Nucleolar-localized protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 307553, 319211, 8715 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O94818, P60954 | |
| Nol4 | |
| Synthesized peptide derived from human NOL4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction