missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | NOL4 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
NOL4 Polyclonal specifically detects NOL4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NOL4 | |
| Polyclonal | |
| Rabbit | |
| O94818-2 | |
| 8715 | |
| Synthetic peptides corresponding to NOL4 (nucleolar protein 4) The peptide sequence was selected from the N terminal of NOL4. Peptide sequence SSNLEERMQSPQNLHGQQDDDSAAESFNGNETLGHSSIASGGTHSREMGD. | |
| Primary |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| CT125, HRIHFB2255, NOLPcancer/testis antigen 125, nucleolar localized protein, nucleolar protein 4, Nucleolar-localized protein | |
| NOL4 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title