missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NOL4 Antibody, Novus Biologicals™

Product Code. 30067042 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30067042 25 μg 25µg
30066976 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30067042 Supplier Novus Biologicals™ Supplier No. NBP33096025ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOL4 Polyclonal antibody specifically detects NOL4 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL4
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot : 1:400 Immunohistochemistry-Paraffin: 2 μg/mL, ELISA, Immunohistochemistry
Formulation Phosphate buffered saline
Gene Alias CT125, HRIHFB2255, NOLPcancer/testis antigen 125, nucleolar localized protein, nucleolar protein 4, Nucleolar-localized protein
Host Species Rabbit
Immunogen Synthetic peptide derived from the human NOL4 protein
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8715
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.