missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NOL4 Polyclonal antibody specifically detects NOL4 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | NOL4 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot : 1:400 Immunohistochemistry-Paraffin: 2 μg/mL, ELISA, Immunohistochemistry |
| Formulation | Phosphate buffered saline |
| Gene Alias | CT125, HRIHFB2255, NOLPcancer/testis antigen 125, nucleolar localized protein, nucleolar protein 4, Nucleolar-localized protein |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from the human NOL4 protein |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?