missing translation for 'onlineSavingsMsg'
Learn More

Mouse Nogo Receptor/NgR Antibody, R&D Systems™
GREENER_CHOICE

Goat Polyclonal Antibody has been used in 12 publications

£184.00 - £463.00

Specifications

Antigen Nogo Receptor/NgR
Concentration LYOPH
Dilution Western Blot 0.1 ug/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18756517
GREENER_CHOICE
View Documents
R&D Systems
AF1440-SP
25 μg
£184.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18011522
GREENER_CHOICE
View Documents
R&D Systems
AF1440
100 μg
£463.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nogo Receptor/NgR Polyclonal specifically detects Nogo Receptor/NgR in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Nogo Receptor/NgR
Western Blot 0.1 ug/mL
Polyclonal
Goat
Mouse
NgR, NgR1, Nogo receptor, Nogo-66 receptor, NogoR, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor, RTN4R
RTN4R
IgG
Affinity Purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
LYOPH
Western Blot
Unconjugated
RUO
Q99PI8
65078
Mouse myeloma cell line NS0-derived recombinant mouse NgR Cys27-Ser447 Accession # Q99PI8
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects mouse Nogo Receptor in direct ELISAs and Western blots. In these formats, approximately 10% cross-reactivity with recombinant human (rh) NgR is observed and less than 1% cross-reactivity with rhNgRH1 and rhNgRH2 is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt