missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Nogo Receptor/NgR Monoclonal antibody specifically detects Nogo Receptor/NgR in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Nogo Receptor/NgR |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | MM0493-6H43 |
| Conjugate | PerCP |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor |
| Host Species | Mouse |
| Immunogen | Human recombinant Nogo Receptor/NgR (extracellular domain) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?