missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nogo Receptor/NgR Antibody (M5), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52675AF647
This item is not returnable.
View return policy
Description
Nogo Receptor/NgR Monoclonal antibody specifically detects Nogo Receptor/NgR in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF
Specifications
| Nogo Receptor/NgR | |
| Monoclonal | |
| Alexa Fluor 647 | |
| NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor | |
| (His)6-tagged fragment of rat NgR1 (amino acids 278-473). | |
| 0.1 mL | |
| Neurodegeneration, Neuroscience | |
| 65078 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF | |
| M5 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction