Learn More
Nogo Receptor/NgR Antibody (M5), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52675AF488
Product Code. 18741471
  • £523.00 / 0.10mL
Description
Nogo Receptor/NgR Monoclonal antibody specifically detects Nogo Receptor/NgR in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nogo Receptor/NgR
Monoclonal
Alexa Fluor 488
NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor
(His)6-tagged fragment of rat NgR1 (amino acids 278-473).
0.1 mL
Neurodegeneration, Neuroscience
65078
Store at 4°C in the dark.
IgG κ
Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF
M5
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified