missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nogo B receptor Rabbit anti-Human, Mouse, Rat, Clone: S01-3K8, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Nogo B receptor Monoclonal antibody specifically detects Nogo B receptor in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Nogo B receptor |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | S01-3K8 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500 |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide |
| Gene Alias | C6orf68MGC117249, chromosome 6 open reading frame 68, MGC:7199, MGC7199, NgBR, nogo-B receptor, Nuclear undecaprenyl pyrophosphate synthase 1 homolog, nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae) |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human Nogo B receptor (Uniprot#: Q96E22) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?