missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Nogo-A Polyclonal specifically detects Nogo-A in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Nogo-A |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50 mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Nogo-A (NP_065393.1; Met1-Val185). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?